siRNA Details
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 4 | 1116 | ||
GenBank mRNA | BC070296.1 | - | 4 | 1095 | ||
BC107767.1 | - | 4 | 1091 |
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 4 | 1120 | ||
GenBank mRNA | AK124365.1 | - | 3 | 2024 | ||
BC005379.1 | - | 4 | 1116 | 3934860 | ||
M93143.2 | - | 4 | 1120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
