siRNA Details
Gene Symbol: RGD1311458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009678.1 | NP_001009678.1 | 6 | 648 | ||
GenBank mRNA | BC088456.1 | - | 6 | 648 | 7127273 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1311458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009678.1 | NP_001009678.1 | 6 | 648 | ||
GenBank mRNA | BC088456.1 | - | 6 | 648 | 7127273 |