siRNA Details
Gene Symbol: MARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317338.1 | NP_001304267.1 | 8 | 2008 | ||
NM_017898.4 | NP_060368.2 | 8 | 2001 | |||
XM_006711407.2 | XP_006711470.1 | 7 | 2557 | |||
XM_011509684.1 | XP_011507986.1 | 8 | 1525 | |||
XM_017001581.1 | XP_016857070.1 | 5 | 1693 | |||
GenBank mRNA | AK000583.1 | - | 1 | 445 | ||
BC010366.1 | - | 7 | 1336 | |||
BC015829.2 | - | 6 | 1271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
