siRNA Details
Gene Symbol: TUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002323.2 | 2 | 3290 | |||
NR_110492.1 | 3 | 3346 | ||||
NR_110493.1 | 2 | 3290 | ||||
GenBank mRNA | AK000749.1 | - | 3 | 1191 | ||
AK057695.1 | - | 1 | 1314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
