siRNA Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010861.2 | NP_001010861.1 | 6 | 3402 | ||
NM_001037317.1 | NP_001032394.1 | 6 | 3417 | |||
XM_011540836.2 | XP_011539138.1 | 7 | 3474 | |||
XM_011540838.2 | XP_011539140.1 | 7 | 3892 | |||
GenBank mRNA | AF131783.1 | - | 1 | 929 | ||
BC040174.1 | - | 6 | 3139 |
