siRNA Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010861.2 | NP_001010861.1 | 6 | 3055 | ||
NM_001037317.1 | NP_001032394.1 | 6 | 3070 | |||
XM_011540836.2 | XP_011539138.1 | 7 | 3127 | |||
XM_011540838.2 | XP_011539140.1 | 7 | 3545 | |||
GenBank mRNA | AF131783.1 | - | 1 | 582 | ||
BC040174.1 | - | 6 | 2792 | |||
BC136350.1 | - | 6 | 2993 | 9020337 | ||
BC144000.1 | - | 6 | 3005 | 9052518 |
