siRNA Details
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010951.1 | NP_001010951.1 | 6, 7 | 854 | ||
XM_006239697.2 | XP_006239759.1 | 7, 8 | 1417 | |||
XM_006239698.2 | XP_006239760.2 | 7, 8 | 982 | |||
XM_006239699.2 | XP_006239761.2 | 7, 8 | 1694 | |||
XM_008764456.1 | XP_008762678.1 | 7, 8 | 1084 | |||
XM_008764457.1 | XP_008762679.1 | 7, 8 | 977 | |||
XM_008764458.1 | XP_008762680.1 | 6, 7 | 777 | |||
GenBank mRNA | BC089111.1 | - | 6, 7 | 854 | 7312962 |

More Information
Other Species Targeted

Gene Ontology Categories:
