siRNA Details
Gene Symbol: TRMT61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017910.3 | NP_060380.3 | 7 | 1453 | ||
XM_005264393.2 | XP_005264450.1 | 7 | 1426 | |||
XM_006712037.2 | XP_006712100.1 | 5 | 1159 | |||
XM_017004401.1 | XP_016859890.1 | 5 | 1132 | |||
XM_017004403.1 | XP_016859892.1 | 7 | 1472 | |||
GenBank mRNA | AK225524.1 | - | 7 | 1445 | ||
AK225551.1 | - | 7 | 1419 | |||
AL136689.1 | - | 7 | 1436 | |||
BC000952.1 | - | 7 | 1443 | 3450078 | ||
BC010365.1 | - | 7 | 1435 | 4284118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
