siRNA Details
Gene Symbol: Olfr324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011743.2 | NP_001011743.2 | 1 | 372 | ||
GenBank mRNA | BC147575.1 | - | 1 | 372 | 9056565 | |
BC147579.1 | - | 1 | 372 | 9056569 |

More Information
Other Species Targeted

Gene Ontology Categories:
