siRNA Details
Gene Symbol: Xkr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012042.1 | NP_001012042.1 | 4 | 2348 | ||
GenBank mRNA | AY534260.1 | - | 4 | 2348 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173393, AY534252.
