siRNA Details
Gene Symbol: FLJ20712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012879.1 | XP_016868368.1 | 1 | 1865 | ||
GenBank mRNA | AK000719.1 | - | 2 | 913 | ||
AK225487.1 | - | 2 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
