siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012068.1 | NP_001012068.1 | 8 | 662 | ||
XM_008759890.1 | XP_008758112.1 | 7 | 863 | |||
XM_008759891.1 | XP_008758113.1 | 7 | 558 | |||
XM_008759892.1 | XP_008758114.1 | 7 | 831 | |||
GenBank mRNA | BC083730.1 | - | 8 | 662 | 7191339 |
