siRNA Details
Gene Symbol: Gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012071.1 | NP_001012071.1 | 5, 6 | 860 | ||
XM_006231590.2 | XP_006231652.1 | 5, 6 | 822 | |||
XM_006231591.2 | XP_006231653.1 | 5, 6 | 794 | |||
XM_006231592.1 | XP_006231654.1 | 5, 6 | 1552 | |||
GenBank mRNA | BC079295.1 | - | 5, 6 | 860 | 7134485 | |
FJ179406.1 | - | 3, 4 | 362 |
Gene Symbol: LOC103690044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760499.1 | XP_008758721.1 | 3, 4 | 1529 |
