siRNA Details
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012087.1 | NP_001012087.1 | 2 | 281 | ||
XM_008761841.1 | XP_008760063.1 | 2 | 972 | |||
XM_008761842.1 | XP_008760064.1 | 1 | 104 | |||
XM_008761843.1 | XP_008760065.1 | 1 | 104 | |||
XM_008761844.1 | XP_008760066.1 | 2 | 562 | |||
GenBank mRNA | BC083842.1 | - | 2 | 281 | 7114399 | |
FQ210426.1 | - | 2 | 255 | |||
FQ222967.1 | - | 2 | 255 | |||
FQ223362.1 | - | 2 | 256 |
