siRNA Details
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012090.1 | NP_001012090.1 | 6 | 822 | ||
GenBank mRNA | BC079122.1 | - | 6 | 822 | 7126707 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012090.1 | NP_001012090.1 | 6 | 822 | ||
GenBank mRNA | BC079122.1 | - | 6 | 822 | 7126707 |