siRNA Details
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012101.2 | NP_001012101.2 | 10, 11 | 1415 | ||
XM_006238058.2 | XP_006238120.1 | 9, 10 | 2970 | |||
GenBank mRNA | BC079209.1 | - | 10, 11 | 1368 | 7131801 |
