siRNA Details
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012101.2 | NP_001012101.2 | 10 | 1373 | ||
XM_006238058.2 | XP_006238120.1 | 9 | 2928 | |||
GenBank mRNA | BC079209.1 | - | 10 | 1326 | 7131801 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001164804, NM_178893, BC031164, BC138732, AK054394, and others.
Gene Ontology Categories:
