siRNA Details
Gene Symbol: Trim32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012103.1 | NP_001012103.1 | 2 | 2873 | ||
XM_006238270.2 | XP_006238332.1 | 2 | 2915 | |||
GenBank mRNA | BC081980.1 | - | 2 | 2873 | 7128474 | |
BC091385.1 | - | 2 | 2833 | 7321183 |
