siRNA Details
Gene Symbol: Plscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012139.2 | NP_001012139.1 | 6 | 842 | ||
XM_008767838.1 | XP_008766060.1 | 5 | 1193 | |||
XM_008767839.1 | XP_008766061.1 | 5 | 2177 | |||
GenBank mRNA | AY548831.1 | - | 5 | 719 | ||
BC098055.1 | - | 6 | 842 | 7457695 |
