siRNA Details
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204817.1 | NP_001191746.1 | 3 | 6344 | ||
NM_032828.3 | NP_116217.1 | 3 | 6347 | |||
GenBank mRNA | AK026676.1 | - | 2 | 1420 | ||
BC017219.2 | - | 2 | 1093 |
Gene Symbol: ZNF417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297734.1 | NP_001284663.1 | 3 | 4101 | ||
NM_152475.2 | NP_689688.2 | 3 | 4098 | |||
XM_011526473.2 | XP_011524775.1 | 3 | 4691 | |||
XM_017026315.1 | XP_016881804.1 | 3 | 3964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
