siRNA Details
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012153.1 | NP_001012153.1 | 19 | 2180 | ||
XM_006251338.2 | XP_006251400.1 | 21 | 2980 | |||
XM_006251339.2 | XP_006251401.1 | 20 | 2947 | |||
GenBank mRNA | BC079058.1 | - | 19 | 2180 | 7115655 | |
FQ229924.1 | - | 20 | 2219 |
