siRNA Details
Gene Symbol: ZNF181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029997.3 | NP_001025168.2 | 4 | 1833 | ||
NM_001145665.1 | NP_001139137.1 | 4 | 1830 | |||
XM_005258849.2 | XP_005258906.1 | 5 | 1601 | |||
XM_005258850.2 | XP_005258907.1 | 5 | 1598 | |||
XM_005258851.3 | XP_005258908.1 | 5 | 1589 | |||
XM_006723183.3 | XP_006723246.1 | 5 | 1855 | |||
XM_006723184.3 | XP_006723247.1 | 5 | 1586 | |||
XM_017026739.1 | XP_016882228.1 | 5 | 2343 | |||
XM_017026740.1 | XP_016882229.1 | 4 | 1463 | |||
GenBank mRNA | AK297316.1 | - | 1 | 665 | ||
BC043228.1 | - | 3 | 2474 | 5295602 | ||
BC104759.1 | - | 4 | 1459 | 8143762 | ||
BC143442.1 | - | 4 | 1456 | 9051950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
