siRNA Details
Gene Symbol: Lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012354.2 | NP_001012354.1 | 1354 | |||
XM_006234777.2 | XP_006234839.1 | 1120 | ||||
XM_006234778.1 | XP_006234840.1 | 1135 | ||||
XM_006234779.2 | XP_006234841.1 | 1150 | ||||
XM_006234780.2 | XP_006234842.1 | 1171 | ||||
GenBank mRNA | BC089966.1 | - | 1171 | 7314989 |
