siRNA Details
Gene Symbol: FLJ10038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026891.1 | 1 | 1046 | |||
GenBank mRNA | AK000900.1 | - | 1 | 1029 | ||
BC009935.2 | - | 1 | 1026 | |||
BC068063.1 | - | 1 | 1019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
