siRNA Details
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012472.1 | NP_001012490.1 | 8, 9 | 1047 | ||
XM_006248635.2 | XP_006248697.1 | 8, 9 | 1092 | |||
XM_006248636.2 | XP_006248698.1 | 7, 8 | 987 | |||
GenBank mRNA | BC089898.1 | - | 8, 9 | 1047 | 7382217 |
