siRNA Details
Gene Symbol: GRAMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012642.2 | NP_001012660.1 | 12 | 2569 | ||
XM_011521327.2 | XP_011519629.1 | 12 | 2678 | |||
XM_011521328.2 | XP_011519630.1 | 12 | 3029 | |||
XM_011521329.2 | XP_011519631.1 | 12 | 2546 | |||
XM_011521330.2 | XP_011519632.1 | 12 | 2630 | |||
XM_011521331.2 | XP_011519633.1 | 11 | 2512 | |||
XM_011521332.2 | XP_011519634.1 | 13 | 2753 | |||
XM_011521333.2 | XP_011519635.1 | 13 | 2770 | |||
XM_017021997.1 | XP_016877486.1 | 12 | 2713 | |||
XM_017021998.1 | XP_016877487.1 | 12 | 2937 | |||
GenBank mRNA | AK002016.1 | - | 1 | 1277 | ||
AK095072.1 | - | 12 | 2559 | |||
AK096501.1 | - | 3 | 2088 | |||
BC008057.2 | - | 1 | 1139 | |||
BC038451.1 | - | 12 | 2543 |
