siRNA Details
Gene Symbol: Pip5k1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012743.1 | NP_001012761.1 | 6 | 899 | ||
XM_008760304.1 | XP_008758526.1 | 6 | 530 | |||
GenBank mRNA | BC090349.1 | - | 6 | 899 | 7311399 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF048695.
Gene Ontology Categories:
