siRNA Details
Gene Symbol: SUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017982.3 | NP_060452.3 | 9 | 2487 | ||
XM_005273169.1 | XP_005273226.1 | 10 | 2510 | |||
XM_005273172.1 | XP_005273229.1 | 10 | 2567 | |||
XM_006711408.1 | XP_006711471.1 | 9 | 2491 | |||
XM_011509685.1 | XP_011507987.1 | 9 | 2525 | |||
XM_011509687.1 | XP_011507989.1 | 10 | 2573 | |||
XM_017001583.1 | XP_016857072.1 | 10 | 2691 | |||
XM_017001584.1 | XP_016857073.1 | 10 | 2684 | |||
XM_017001585.1 | XP_016857074.1 | 11 | 2674 | |||
XM_017001586.1 | XP_016857075.1 | 10 | 2568 | |||
XM_017001587.1 | XP_016857076.1 | 10 | 2504 | |||
XM_017001588.1 | XP_016857077.1 | 9 | 2519 | |||
GenBank mRNA | AK000914.1 | - | 7 | 1692 | ||
AK096265.1 | - | 8 | 2941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
