siRNA Details
Gene Symbol: Cstf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013161.1 | NP_001013179.1 | 6 | 1652 | ||
XM_006235681.1 | XP_006235743.1 | 6 | 1757 | |||
XM_006235682.1 | XP_006235744.1 | 6 | 1701 | |||
XM_006235683.2 | XP_006235745.1 | 6 | 1784 | |||
XM_006235684.2 | XP_006235746.1 | 6 | 1781 | |||
XM_006235685.2 | XP_006235747.1 | 6 | 1759 | |||
XM_006235686.2 | XP_006235748.1 | 6 | 1756 | |||
XM_006235687.2 | XP_006235749.1 | 6 | 1753 | |||
GenBank mRNA | BC091381.1 | - | 6 | 1652 | 7320917 |
