siRNA Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013170.2 | NP_001013188.2 | 2, 3 | 231 | ||
XM_006240542.2 | XP_006240604.1 | 2, 3 | 268 | |||
XM_008764811.1 | XP_008763033.1 | 2, 3 | 422 | |||
XM_008764812.1 | XP_008763034.1 | 2, 3 | 1019 | |||
GenBank mRNA | AY724504.1 | - | 2, 3 | 231 |
