siRNA Details
Gene Symbol: LOC441233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128010.1 | - | 1 | 1924 |
Gene Symbol: LOC100240728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108095.1 | 2 | 661 | |||
GenBank mRNA | BC036258.1 | - | 2 | 637 |
