siRNA Details
Gene Symbol: LOC298139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013930.1 | NP_001013952.1 | 2 | 1821 | ||
GenBank mRNA | BC083698.1 | - | 2 | 1821 | 7189881 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC298139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013930.1 | NP_001013952.1 | 2 | 1821 | ||
GenBank mRNA | BC083698.1 | - | 2 | 1821 | 7189881 |