siRNA Details
Gene Symbol: Fam214b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013931.1 | NP_001013953.1 | 9 | 2005 | ||
GenBank mRNA | BC087107.1 | - | 9 | 2005 | 7193207 | |
FQ212427.1 | - | 2023 | ||||
FQ212481.1 | - | 2023 |
