siRNA Details
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013995.1 | NP_001014017.1 | 5, 6 | 642 | ||
XM_006251955.1 | XP_006252017.1 | 5, 6 | 614 | |||
XM_006251956.2 | XP_006252018.1 | 5, 6 | 793 | |||
GenBank mRNA | BC079109.1 | - | 5, 6 | 642 | 7125564 |
