siRNA Details
Gene Symbol: Pde12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013998.2 | NP_001014020.2 | 3 | 1323 | ||
GenBank mRNA | AY387060.1 | - | 3 | 1323 | ||
BC079396.1 | - | 3 | 1507 | 7112419 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pde12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013998.2 | NP_001014020.2 | 3 | 1323 | ||
GenBank mRNA | AY387060.1 | - | 3 | 1323 | ||
BC079396.1 | - | 3 | 1507 | 7112419 |