siRNA Details
Gene Symbol: Gramd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014011.1 | NP_001014033.1 | 14 | 2133 | ||
XM_006254748.2 | XP_006254810.1 | 14 | 2235 | |||
XM_008772140.1 | XP_008770362.1 | 14 | 2060 | |||
GenBank mRNA | BC090001.1 | - | 14 | 2133 | 7321780 |
