siRNA Details
Gene Symbol: Nip30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014014.1 | NP_001014036.1 | 8 | 1230 | ||
XM_006255110.2 | XP_006255172.1 | 8 | 1540 | |||
XM_006255111.2 | XP_006255173.1 | 8 | 1307 | |||
XM_006255114.2 | XP_006255176.1 | 9 | 1772 | |||
GenBank mRNA | BC079145.1 | - | 8 | 1230 | 7128234 |
