siRNA Details
Gene Symbol: Zfp819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014017.2 | NP_001014039.1 | 5 | 1836 | ||
XM_006229044.2 | XP_006229106.1 | 6 | 1853 | |||
XM_006229045.2 | XP_006229107.1 | 7 | 3809 | |||
XM_006229046.2 | XP_006229108.1 | 5 | 1793 | |||
XM_006229047.2 | XP_006229109.1 | 5 | 1911 | |||
XM_008759347.1 | XP_008757569.1 | 7 | 2991 | |||
XM_008759348.1 | XP_008757570.1 | 8 | 8604 | |||
XM_008759349.1 | XP_008757571.1 | 6 | 4277 | |||
XM_008759350.1 | XP_008757572.1 | 4 | 1587 | |||
GenBank mRNA | BC087070.1 | - | 5 | 1840 | 7134052 |
