siRNA Details
Gene Symbol: Zfp819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014017.2 | NP_001014039.1 | 5 | 2247 | ||
XM_006229044.2 | XP_006229106.1 | 6 | 2264 | |||
XM_006229045.2 | XP_006229107.1 | 7 | 4220 | |||
XM_006229046.2 | XP_006229108.1 | 5 | 2204 | |||
XM_006229047.2 | XP_006229109.1 | 5 | 2322 | |||
XM_008759347.1 | XP_008757569.1 | 7 | 3402 | |||
XM_008759348.1 | XP_008757570.1 | 8 | 9015 | |||
XM_008759349.1 | XP_008757571.1 | 6 | 4688 | |||
XM_008759350.1 | XP_008757572.1 | 4 | 1998 | |||
GenBank mRNA | BC087070.1 | - | 5 | 2251 | 7134052 |
