siRNA Details
Gene Symbol: RBM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018047.2 | NP_060517.1 | 3, 4 | 259 | ||
GenBank mRNA | AK001152.1 | - | 3, 4 | 214 | ||
AL136933.1 | - | 3, 4 | 188 | |||
BC003402.1 | - | 3, 4 | 192 | 3449258 | ||
DA534416.1 | - | 3, 4 | 260 |
