siRNA Details
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014027.1 | NP_001014049.1 | 9 | 1498 | ||
XM_006230557.2 | XP_006230619.1 | 11 | 1837 | |||
XM_008759993.1 | XP_008758215.1 | 10 | 1804 | |||
XM_008759994.1 | XP_008758216.1 | 10 | 1810 | |||
XM_008759995.1 | XP_008758217.1 | 10 | 1762 | |||
XM_008759996.1 | XP_008758218.1 | 10 | 2365 | |||
GenBank mRNA | BC090010.1 | - | 9 | 1498 | 7324394 |
Gene Symbol: LOC100911440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749008.3 | XP_003749056.1 | 1837 | |||
XM_008760026.1 | XP_008758248.1 | 1804 | ||||
XM_008760027.1 | XP_008758249.1 | 1810 | ||||
XM_008760028.1 | XP_008758250.1 | 1762 | ||||
XM_008760029.1 | XP_008758251.1 | 2365 |

More Information
Other Species Targeted

Gene Ontology Categories:
