siRNA Details
Gene Symbol: Arl14ep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014045.1 | NP_001014067.1 | 5 | 1227 | ||
XM_008762092.1 | XP_008760314.1 | 5 | 2608 | |||
GenBank mRNA | BC089920.1 | - | 5 | 1227 | 7306710 | |
FQ216376.1 | - | 4 | 815 |
