siRNA Details
Gene Symbol: Pars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014064.1 | NP_001014086.1 | 2 | 1472 | ||
XM_006238512.2 | XP_006238574.1 | 2 | 1511 | |||
XM_006238513.2 | XP_006238575.1 | 2 | 1322 | |||
GenBank mRNA | BC088403.1 | - | 2 | 1472 | 7107369 |

More Information
Other Species Targeted

Gene Ontology Categories:
