siRNA Details
Gene Symbol: RGD1307947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014083.2 | NP_001014105.2 | 7 | 2028 | ||
GenBank mRNA | BC079461.1 | - | 7 | 1996 | 7122037 | |
FQ231099.1 | - | 1 | 866 |
