siRNA Details
Gene Symbol: Plscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014094.1 | NP_001014116.1 | 4 | 413 | ||
XM_008766450.1 | XP_008764672.1 | 4 | 660 | |||
GenBank mRNA | BC079090.1 | - | 4 | 413 | 7123519 |
