siRNA Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014119.2 | NP_001014141.2 | 3 | 514 | ||
XM_006247045.2 | XP_006247107.1 | 3 | 635 | |||
XM_008768069.1 | XP_008766291.1 | 3 | 620 | |||
GenBank mRNA | BC088444.1 | - | 3 | 508 | 7126427 | |
DY311021.1 | - | 3 | 514 |
