siRNA Details
Gene Symbol: Abcg3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014133.1 | NP_001014155.1 | 1480 | |||
GenBank mRNA | BC085773.1 | - | 1480 | 7096429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
