siRNA Details
Gene Symbol: Rab30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015012.1 | NP_001015012.1 | 3 | 1162 | ||
XM_006229692.2 | XP_006229754.1 | 5 | 1387 | |||
XM_006229693.2 | XP_006229755.1 | 5 | 1356 | |||
XM_008759647.1 | XP_008757869.1 | 4 | 4354 | |||
GenBank mRNA | BC091182.1 | - | 3 | 1162 | 7326213 | |
FQ210359.1 | - | 4 | 1263 |
