siRNA Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017374.2 | NP_001017374.2 | 16 | 1849 | ||
GenBank mRNA | AB256044.1 | - | 2 | 76 | ||
AM258959.1 | - | 2 | 76 | |||
BC091330.1 | - | 2 | 160 | |||
BC099117.1 | - | 16 | 1736 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 1671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
