siRNA Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017374.2 | NP_001017374.2 | 22 | 3180 | ||
GenBank mRNA | AB256044.1 | - | 8 | 1407 | ||
AM258959.1 | - | 8 | 1407 | |||
BC091330.1 | - | 8 | 1491 | |||
BC099117.1 | - | 22 | 3067 | |||
FQ227956.1 | - | 2 | 345 | |||
FQ232649.1 | - | 5 | 812 |

More Information
Other Species Targeted

Gene Ontology Categories:
